Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600003452.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003452.mm.1 |
NM_001145952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003452.mm.1 |
NM_001145952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003452.mm.1 |
NM_178665.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003452.mm.1 |
NM_178665.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003449.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003449.mm.1 |
NM_001145952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003449.mm.1 |
NM_001145952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003451.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003451.mm.1 |
NM_001145952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003451.mm.1 |
NM_001145952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003451.mm.1 |
NM_178665.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003451.mm.1 |
NM_178665.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003448.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003448.mm.1 |
NM_001145952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003448.mm.1 |
NM_001145952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003445.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003445.mm.1 |
NM_178665.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003445.mm.1 |
NM_178665.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003450.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003450.mm.1 |
NM_001145952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003450.mm.1 |
NM_001145952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003450.mm.1 |
NM_178665.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003450.mm.1 |
NM_178665.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003447.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003447.mm.1 |
NM_178665.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003447.mm.1 |
NM_178665.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003454.mm.1 |
TC1600000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003454.mm.1 |
NM_001145952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003454.mm.1 |
NM_001145952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003454.mm.1 |
NM_178665.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003454.mm.1 |
NM_178665.5 |
Refseq |