Assay Details
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027150.1 | NP_081426.1 | 1 - 2 | 174 | 77 | |
GenBank mRNA | AK009836.1 | - | 1 - 2 | 98 | 77 | |
BC089626.1 | - | 1 - 2 | 144 | 6757304 | 77 | |
BC131903.1 | - | 1 - 2 | 174 | 40130180 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
