Assay Details
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011840.2 | NP_035970.1 | 11 - 12 | 1330 | 154 | |
GenBank mRNA | AB019374.1 | - | 11 - 12 | 1331 | 154 | |
AK013856.1 | - | 3 - 4 | 173 | 154 | ||
AK159261.1 | - | 11 - 12 | 1380 | 153 | ||
BC028260.1 | - | 11 - 12 | 1210 | 5067818 | 154 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK013856.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
