Assay Details
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011840.2 | NP_035970.1 | 16 - 17 | 1641 | 94 | |
GenBank mRNA | AB019374.1 | - | 16 - 17 | 1642 | 94 | |
AK013856.1 | - | 8 - 9 | 484 | 94 | ||
AK159261.1 | - | 16 - 17 | 1691 | 94 | ||
AK178825.1 | - | 1 - 2 | 33 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
