Assay Details
Gene Symbol: Dennd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162917.1 | NP_001156389.1 | 28 - 29 | 5313 | 58 | |
GenBank mRNA | AK035923.1 | - | 6 - 7 | 1411 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
