Assay Details
Gene Symbol: Dennd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162917.1 | NP_001156389.1 | 30 - 31 | 5695 | 77 | |
GenBank mRNA | AK035923.1 | - | 8 - 9 | 1793 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
