Assay Details
Gene Symbol: Zwilch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026507.4 | NP_080783.3 | 10 - 11 | 1034 | 149 | |
GenBank mRNA | AK009559.1 | - | 10 - 11 | 1005 | 149 | |
AK019825.1 | - | 10 - 11 | 1012 | 149 | ||
AK050917.1 | - | 10 - 11 | 1000 | 149 | ||
BC027435.1 | - | 10 - 11 | 982 | 2812472 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
