Assay Details
Gene Symbol: Eef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007907.2 | NP_031933.1 | 2 - 3 | 316 | 62 | |
GenBank mRNA | AK028938.1 | - | 2 - 3 | 295 | 62 | |
AK032997.1 | - | 2 - 3 | 302 | 62 | ||
AK040474.1 | - | 2 - 3 | 302 | 62 | ||
AK077866.1 | - | 2 - 3 | 302 | 62 | ||
AK087985.1 | - | 2 - 3 | 302 | 62 | ||
AK128976.1 | - | 2 - 3 | 301 | 62 | ||
AK134195.1 | - | 2 - 3 | 302 | 62 | ||
AK144874.1 | - | 2 - 3 | 304 | 62 | ||
AK145509.1 | - | 2 - 3 | 302 | 62 | ||
AK145545.1 | - | 2 - 3 | 302 | 62 | ||
AK146508.1 | - | 2 - 3 | 302 | 62 | ||
AK146947.1 | - | 2 - 3 | 302 | 62 | ||
AK149659.1 | - | 2 - 3 | 302 | 62 | ||
AK150136.1 | - | 2 - 3 | 302 | 62 | ||
AK150902.1 | - | 2 - 3 | 302 | 62 | ||
AK151487.1 | - | 2 - 3 | 302 | 62 | ||
AK151812.1 | - | 2 - 3 | 252 | 62 | ||
AK151945.1 | - | 2 - 3 | 302 | 62 | ||
AK152146.1 | - | 2 - 3 | 302 | 62 | ||
AK152447.1 | - | 2 - 3 | 302 | 62 | ||
AK152587.1 | - | 2 - 3 | 302 | 62 | ||
AK152826.1 | - | 2 - 3 | 302 | 62 | ||
AK153275.1 | - | 2 - 3 | 302 | 62 | ||
AK153936.1 | - | 2 - 3 | 302 | 62 | ||
AK155325.1 | - | 2 - 3 | 302 | 62 | ||
AK159155.1 | - | 2 - 3 | 302 | 62 | ||
AK159421.1 | - | 2 - 3 | 302 | 62 | ||
AK159514.1 | - | 2 - 3 | 302 | 62 | ||
AK159806.1 | - | 2 - 3 | 302 | 62 | ||
AK159829.1 | - | 2 - 3 | 302 | 62 | ||
AK163063.1 | - | 2 - 3 | 302 | 62 | ||
AK166212.1 | - | 2 - 3 | 302 | 62 | ||
AK166595.1 | - | 2 - 3 | 302 | 62 | ||
AK166596.1 | - | 2 - 3 | 302 | 62 | ||
AK166719.1 | - | 2 - 3 | 302 | 62 | ||
AK166747.1 | - | 2 - 3 | 302 | 62 | ||
AK166751.1 | - | 2 - 3 | 300 | 62 | ||
AK166758.1 | - | 2 - 3 | 302 | 62 | ||
AK166788.1 | - | 2 - 3 | 302 | 62 | ||
AK166815.1 | - | 2 - 3 | 302 | 62 | ||
AK166829.1 | - | 2 - 3 | 302 | 62 | ||
AK166851.1 | - | 2 - 3 | 300 | 62 | ||
AK166968.1 | - | 2 - 3 | 302 | 62 | ||
AK167081.1 | - | 2 - 3 | 302 | 62 | ||
AK167093.1 | - | 2 - 3 | 302 | 62 | ||
AK167109.1 | - | 2 - 3 | 302 | 62 | ||
AK167115.1 | - | 2 - 3 | 302 | 62 | ||
AK167199.1 | - | 2 - 3 | 302 | 62 | ||
AK167208.1 | - | 2 - 3 | 300 | 62 | ||
AK167221.1 | - | 2 - 3 | 302 | 62 | ||
AK167230.1 | - | 2 - 3 | 302 | 62 | ||
AK167281.1 | - | 2 - 3 | 301 | 62 | ||
AK168527.1 | - | 2 - 3 | 302 | 62 | ||
AK168615.1 | - | 2 - 3 | 302 | 62 | ||
AK168638.1 | - | 2 - 3 | 302 | 62 | ||
AK168659.1 | - | 2 - 3 | 302 | 62 | ||
AK168794.1 | - | 2 - 3 | 302 | 62 | ||
AK168827.1 | - | 2 - 3 | 302 | 62 | ||
AK168874.1 | - | 2 - 3 | 295 | 62 | ||
AK169013.1 | - | 2 - 3 | 302 | 62 | ||
AK189260.1 | - | 1 - 2 | 163 | 62 | ||
AK211738.1 | - | 1 - 2 | 163 | 62 | ||
AK212666.1 | - | 1 - 2 | 163 | 62 | ||
AK214266.1 | - | 1 - 2 | 227 | 62 | ||
AK215787.1 | - | 1 - 2 | 163 | 62 | ||
BC007152.1 | - | 2 - 3 | 300 | 3600352 | 62 | |
BC013263.1 | - | 2 - 3 | 288 | 62 | ||
BC060707.1 | - | 1 - 2 | 177 | 62 | ||
CJ170931.1 | - | 2 - 3 | 317 | 62 | ||
U89415.1 | - | 1 - 2 | 197 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
