Assay Details
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007399.3 | NP_031425.2 | 9 - 10 | 1383 | 134 | |
GenBank mRNA | AF011379.1 | - | 9 - 10 | 1204 | 134 | |
AK036599.1 | - | 3 - 4 | 427 | 134 | ||
AK036883.1 | - | 1 - 2 | 71 | 134 | ||
AK036918.1 | - | 1 - 2 | 72 | 134 | ||
BC057346.1 | - | 9 - 10 | 1344 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
