Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 10 - 11 | 1815 | 108 | |
NM_001110351.1 | NP_001103821.1 | 11 - 12 | 1692 | 108 | ||
NM_011378.2 | NP_035508.2 | 11 - 12 | 1923 | 108 | ||
GenBank mRNA | AK032209.1 | - | 10 - 11 | 1683 | 108 | |
AK136347.1 | - | 5 - 6 | 617 | 108 | ||
AK220292.1 | - | 10 - 11 | 1792 | 108 | ||
BC052716.1 | - | 10 - 11 | 1702 | 6837144 | 108 | |
BC053385.1 | - | 10 - 11 | 1685 | 108 | ||
BC078454.1 | - | 10 - 11 | 1676 | 108 | ||
L36831.1 | - | 10 - 11 | 1606 | 108 | ||
L38620.2 | - | 9 - 10 | 1582 | 108 | ||
L38621.2 | - | 9 - 10 | 1582 | 108 | ||
U22394.1 | - | 11 - 12 | 1931 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
