Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 12 - 13 | 2140 | 121 | |
NM_001110351.1 | NP_001103821.1 | 13 - 14 | 2017 | 121 | ||
NM_011378.2 | NP_035508.2 | 13 - 14 | 2248 | 121 | ||
GenBank mRNA | AK032209.1 | - | 12 - 13 | 2008 | 121 | |
AK136347.1 | - | 7 - 8 | 942 | 121 | ||
AK220292.1 | - | 12 - 13 | 2117 | 121 | ||
BC052716.1 | - | 12 - 13 | 2027 | 6837144 | 121 | |
BC053385.1 | - | 12 - 13 | 2010 | 121 | ||
BC078454.1 | - | 12 - 13 | 2001 | 121 | ||
L36831.1 | - | 12 - 13 | 1931 | 121 | ||
L38620.2 | - | 11 - 12 | 1907 | 121 | ||
L38621.2 | - | 11 - 12 | 1907 | 121 | ||
U22394.1 | - | 13 - 14 | 2256 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
