Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 5 - 6 | 1042 | 174 | |
NM_001110351.1 | NP_001103821.1 | 6 - 7 | 919 | 174 | ||
NM_011378.2 | NP_035508.2 | 6 - 7 | 1150 | 174 | ||
GenBank mRNA | AK032209.1 | - | 5 - 6 | 910 | 174 | |
AK220292.1 | - | 5 - 6 | 1019 | 174 | ||
BC052716.1 | - | 5 - 6 | 929 | 6837144 | 174 | |
BC053385.1 | - | 5 - 6 | 912 | 174 | ||
BC078454.1 | - | 5 - 6 | 903 | 174 | ||
L36831.1 | - | 5 - 6 | 833 | 174 | ||
L38620.2 | - | 4 - 5 | 809 | 174 | ||
L38621.2 | - | 4 - 5 | 809 | 174 | ||
U22394.1 | - | 6 - 7 | 1158 | 174 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
