Assay Details
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010301.3 | NP_034431.1 | 6 - 7 | 1011 | 96 | |
GenBank mRNA | AK139289.1 | - | 2 - 3 | 850 | 96 | |
AK143685.1 | - | 6 - 7 | 1012 | 96 | ||
BC011169.1 | - | 6 - 7 | 1010 | 4206878 | 96 | |
M55411.1 | - | 6 - 7 | 948 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
