Assay Details
Gene Symbol: Lss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031271.1 | - | 9 - 10 | 1061 | 80 | |
AK054393.1 | - | 9 - 10 | 1073 | 80 | ||
AK078023.1 | - | 9 - 10 | 1054 | 80 | ||
AK141216.1 | - | 9 - 10 | 1073 | 80 | ||
AK147680.1 | - | 9 - 10 | 1737 | 80 | ||
AK154520.1 | - | 9 - 10 | 1067 | 80 | ||
BC029082.1 | - | 9 - 10 | 1051 | 3498101 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
