Assay Details
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB093224.1 | - | 10 - 11 | 3093 | 68 | |
AF363690.1 | - | 15 - 16 | 3826 | 68 | ||
AK004685.1 | - | 6 - 7 | 1219 | 68 | ||
AK082152.1 | - | 5 - 6 | 983 | 68 | ||
AK147276.1 | - | 15 - 16 | 4453 | 68 | ||
AK147535.1 | - | 16 - 17 | 6571 | 68 | ||
BC040463.1 | - | 6 - 7 | 1520 | 68 | ||
BC058665.1 | - | 12 - 13 | 3338 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
