Assay Details
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143776.1 | NP_001137248.1 | 8 - 9 | 1109 | 116 | |
NM_001143777.1 | NP_001137249.1 | 9 - 10 | 1213 | 116 | ||
NM_024244.4 | NP_077206.3 | 8 - 9 | 1109 | 116 | ||
GenBank mRNA | AK004797.1 | - | 8 - 9 | 1093 | 116 | |
AK032053.1 | - | 9 - 10 | 1025 | 116 | ||
AK036299.1 | - | 9 - 10 | 1176 | 116 | ||
AK041147.1 | - | 8 - 9 | 1110 | 116 | ||
AK044049.1 | - | 9 - 10 | 1136 | 116 | ||
AK082202.1 | - | 8 - 9 | 1105 | 116 | ||
AK220421.1 | - | 9 - 10 | 1049 | 116 | ||
BC129839.1 | - | 8 - 9 | 1065 | 40060810 | 116 | |
BC129840.1 | - | 8 - 9 | 1065 | 40060811 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
