Assay Details
Gene Symbol: Tpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164255.1 | NP_001157727.1 | 7 - 8 | 787 | 124 | |
NM_001164256.1 | NP_001157728.1 | 6 - 7 | 760 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
