Assay Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081346.1 | NP_001074815.1 | 3 - 4 | 468 | 105 | |
GenBank mRNA | AK030986.1 | - | 3 - 4 | 352 | 105 | |
AK045134.1 | - | 369 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
