Assay Details
Gene Symbol: Gm20611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040638.1 | 1 - 2 | 144 | 85 | ||
GenBank mRNA | AK029795.1 | - | 1 - 2 | 144 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 26947 |
Gm26947 | -90.4kb | Sense | Chr. 10 - 60931986 - 60940944 on NCBI Build GRCm38 | |
Downstream | pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 1 | Pcbd1 |
Dcoh, Pcbd, Pcd, Phs |
52.2kb | Sense | Chr. 10 - 61089331 - 61094329 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
