Assay Details
Gene Symbol: Kirrel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172898.3 | NP_766486.1 | 14 - 15 | 2427 | 74 | |
GenBank mRNA | AK049284.1 | - | 14 - 15 | 2428 | 74 | |
AK083069.1 | - | 12 - 13 | 2977 | 74 | ||
BC052773.1 | - | 14 - 15 | 1890 | 6314924 | 74 | |
BC158021.1 | - | 14 - 15 | 1935 | 9007327 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031609 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031590 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031591 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031610 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031611 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031593 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | BC052773.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | AK083069.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | NM_172898.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | AK049284.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | BC052773.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | AK083069.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | NM_172898.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512306 | AK049284.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154005 | NM_172898.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154005 | AK049284.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973268 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 60304 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449888 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874283 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454526 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122667 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111588 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177636 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154005 | NM_172898.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154005 | AK049284.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | BC052773.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | AK083069.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | NM_172898.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | AK049284.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | BC052773.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | AK083069.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | NM_172898.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512306 | AK049284.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031576 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031579 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031578 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031596 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031611 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031581 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031629 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25031622 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177636 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06154598 | - | - |
Whole Mouse Genome G4122A | A_52_P539095 | BC052773.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P539095 | BC158021.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P539095 | AK083069.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P539095 | AY305303.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P539095 | NM_172898.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | BC052773.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | AK083069.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | NM_172898.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | AK049284.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | BC052773.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | AK083069.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | NM_172898.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P512306 | AK049284.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105842 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177636 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06228029 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06228032 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06228026 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06228018 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06102497 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06151355 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06102500 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06228021 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06102491 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06151362 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06151361 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06102492 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06228016 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031591 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031593 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031609 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031590 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031611 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25031610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06348851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06660211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06144932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06382544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031585 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06144930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031616 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06228026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06144926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06660213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25031607 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10561948 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561951 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561945 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561953 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561952 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561950 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561946 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561954 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561943 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561944 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561947 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10561949 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10561942 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700024481.mm.1 | TC0700002760.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700024481.mm.1 | NM_172898.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700024481.mm.1 | NM_172898.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700024479.mm.1 | TC0700002760.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700024479.mm.1 | NM_172898.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700024479.mm.1 | NM_172898.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700024482.mm.1 | TC0700002760.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700024482.mm.1 | NM_172898.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700024482.mm.1 | NM_172898.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700024478.mm.1 | TC0700002760.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 558922 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 130168 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 130168 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3440097 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000130168 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003440097 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2629246 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608247 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2608247 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2629246 | - | - |
mouse-6 v1.1 (Illumina) | scl31525.11.1_99-S | - | - |
mouse-6 v1.1 (Illumina) | scl00074.1_47-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3440097 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 130168 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl31525.11.1_99-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00074.1_47-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440097 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130168 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2629246 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2629246 | - | - |
