Assay Details
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020274.4 | NP_064670.1 | 1 - 2 | 237 | 77 | |
GenBank mRNA | AF155045.1 | - | 1 - 2 | 69 | 77 | |
AK085213.1 | - | 1 - 2 | 238 | 77 | ||
BC046990.1 | - | 1 - 2 | 215 | 6309747 | 77 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF155045.1, AK085213.1, BC046990.1, NM_020274.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
