Assay Details
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020274.4 | NP_064670.1 | 5 - 6 | 724 | 65 | |
GenBank mRNA | AF155045.1 | - | 5 - 6 | 556 | 65 | |
AK085213.1 | - | 5 - 6 | 725 | 65 | ||
BC046990.1 | - | 5 - 6 | 702 | 6309747 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
