Assay Details
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020274.4 | NP_064670.1 | 6 - 7 | 889 | 62 | |
GenBank mRNA | AF155045.1 | - | 6 - 7 | 721 | 62 | |
AK085213.1 | - | 6 - 7 | 890 | 62 | ||
BC046990.1 | - | 6 - 7 | 867 | 6309747 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
