Assay Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099644.1 | NP_001093114.1 | 3 - 4 | 411 | 83 | |
NM_013561.2 | NP_038589.2 | 3 - 4 | 411 | 83 | ||
GenBank mRNA | AF513221.1 | - | 3 - 4 | 288 | 83 | |
AY605711.1 | - | 3 - 4 | 277 | 83 | ||
BC144799.1 | - | 3 - 4 | 333 | 9053347 | 83 | |
BC145743.1 | - | 3 - 4 | 333 | 40131046 | 83 | |
M74425.1 | - | 3 - 4 | 334 | 83 | ||
X72395.1 | - | 3 - 4 | 295 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
