Assay Details
Gene Symbol: Npat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081152.1 | NP_001074621.1 | 11 - 12 | 1111 | 95 | |
GenBank mRNA | AK033012.1 | - | 11 - 12 | 1091 | 95 | |
AK080294.1 | - | 11 - 12 | 1112 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
