Assay Details
Gene Symbol: Chrnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148944.4 | NP_683746.1 | 1 - 2 | 100 | 90 | |
GenBank mRNA | AF492840.1 | - | 1 - 2 | 85 | 90 | |
AK140071.1 | - | 1 - 2 | 102 | 90 | ||
AY574269.1 | - | 1 - 2 | 51 | 90 | ||
BC094889.1 | - | 1 - 2 | 146 | 90 | ||
BC138851.1 | - | 1 - 2 | 99 | 8861873 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
