Assay Details
Gene Symbol: Ireb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022655.3 | NP_073146.2 | 16 - 17 | 2194 | 123 | |
GenBank mRNA | BC044665.1 | - | 16 - 17 | 2161 | 2647641 | 123 |
CV558274.1 | - | 4 - 5 | 476 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
