Assay Details
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172787.2 | NP_766375.1 | 17 - 18 | 1829 | 120 | |
GenBank mRNA | AK030412.1 | - | 4 - 5 | 427 | 120 | |
AK045667.1 | - | 17 - 18 | 1830 | 120 | ||
AK049597.1 | - | 17 - 18 | 1864 | 120 | ||
AK129443.1 | - | 13 - 14 | 1374 | 120 | ||
BC082309.1 | - | 16 - 17 | 1699 | 30356672 | 120 | |
BC085192.1 | - | 16 - 17 | 1934 | 30694487 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
