Assay Details
Gene Symbol: P3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042411.1 | NP_001035874.1 | 6 - 7 | 854 | 102 | |
NM_001286148.1 | NP_001273077.1 | 6 - 7 | 1220 | 102 | ||
NM_019782.3 | NP_062756.2 | 6 - 7 | 1220 | 102 | ||
NM_019783.2 | NP_062757.2 | 6 - 7 | 1220 | 102 | ||
GenBank mRNA | AK010578.1 | - | 6 - 7 | 1222 | 102 | |
AK030436.1 | - | 6 - 7 | 1221 | 102 | ||
AK159505.1 | - | 6 - 7 | 855 | 102 | ||
BC024047.1 | - | 6 - 7 | 1186 | 5148677 | 102 | |
CX563255.1 | - | 2 - 3 | 112 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
