Assay Details
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172495.5 | NP_766083.3 | 11 - 12 | 2588 | 62 | |
GenBank mRNA | AK128935.1 | - | 10 - 11 | 2411 | 62 | |
BC042791.1 | - | 668 | 62 | |||
BC076623.1 | - | 9 - 10 | 2194 | 62 | ||
BC141150.1 | - | 10 - 11 | 2289 | 9055762 | 62 | |
BC145459.1 | - | 9 - 10 | 2174 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
