Assay Details
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111267.2 | NP_001104737.1 | 4 - 5 | 613 | 113 | |
NM_172495.5 | NP_766083.3 | 14 - 15 | 2964 | 113 | ||
GenBank mRNA | AK033503.1 | - | 4 - 5 | 527 | 113 | |
AK042369.1 | - | 4 - 5 | 583 | 113 | ||
AK128935.1 | - | 13 - 14 | 2787 | 113 | ||
BC042791.1 | - | 1044 | 113 | |||
BC049997.1 | - | 4 - 5 | 563 | 113 | ||
BC058572.1 | - | 4 - 5 | 535 | 113 | ||
BC076623.1 | - | 12 - 13 | 2570 | 113 | ||
BC080742.1 | - | 3 - 4 | 1719 | 113 | ||
BC141150.1 | - | 13 - 14 | 2665 | 9055762 | 113 | |
BC145459.1 | - | 12 - 13 | 2550 | 113 | ||
EF602028.1 | - | 4 - 5 | 613 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
