Assay Details
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172495.5 | NP_766083.3 | 7 - 8 | 1054 | 102 | |
GenBank mRNA | AK128935.1 | - | 6 - 7 | 877 | 102 | |
BC076623.1 | - | 5 - 6 | 660 | 102 | ||
BC141150.1 | - | 6 - 7 | 788 | 9055762 | 102 | |
BC145459.1 | - | 5 - 6 | 674 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
