Assay Details
Gene Symbol: Ephb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290753.1 | NP_001277682.1 | 13 - 14 | 2681 | 66 | |
NM_010142.3 | NP_034272.1 | 13 - 14 | 2678 | 66 | ||
GenBank mRNA | BC043088.1 | - | 13 - 14 | 2602 | 6418088 | 66 |
BC062924.1 | - | 13 - 14 | 2626 | 66 | ||
L25890.1 | - | 14 - 15 | 2520 | 66 | ||
X76011.1 | - | 7 - 8 | 979 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
