Assay Details
Gene Symbol: Ephb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290753.1 | NP_001277682.1 | 6 - 7 | 1614 | 58 | |
GenBank mRNA | BC062924.1 | - | 6 - 7 | 1559 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
