Assay Details
Gene Symbol: Kdm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133872.2 | NP_598633.2 | 1 - 2 | 490 | 99 | |
GenBank mRNA | AK129170.1 | - | 1 - 2 | 430 | 99 | |
BC059885.1 | - | 1 - 2 | 477 | 6815975 | 99 | |
CJ134496.1 | - | 1 - 2 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
