Assay Details
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111268.1 | NP_001104738.1 | 14 - 15 | 2495 | 123 | |
NM_010349.2 | NP_034479.2 | 14 - 15 | 2495 | 123 | ||
GenBank mRNA | BC131639.1 | - | 13 - 14 | 2176 | 123 | |
BC131640.1 | - | 13 - 14 | 2176 | 123 | ||
BC148311.2 | - | 13 - 14 | 2176 | 123 | ||
D10054.1 | - | 14 - 15 | 2408 | 123 | ||
X66117.1 | - | 13 - 14 | 2232 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
