Assay Details
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111268.1 | NP_001104738.1 | 1 - 2 | 126 | 82 | |
NM_010349.2 | NP_034479.2 | 1 - 2 | 126 | 82 | ||
GenBank mRNA | AK141539.1 | - | 1 - 2 | 127 | 82 | |
D10054.1 | - | 1 - 2 | 39 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK141539.1, D10054.1, NM_001111268.1, NM_010349.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
