Assay Details
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111268.1 | NP_001104738.1 | 3 - 4 | 697 | 107 | |
NM_010349.2 | NP_034479.2 | 3 - 4 | 697 | 107 | ||
GenBank mRNA | AK141539.1 | - | 3 - 4 | 698 | 107 | |
BC060154.1 | - | 2 - 3 | 492 | 107 | ||
BC131639.1 | - | 2 - 3 | 378 | 107 | ||
BC131640.1 | - | 2 - 3 | 378 | 107 | ||
BC148311.2 | - | 2 - 3 | 378 | 107 | ||
D10054.1 | - | 3 - 4 | 610 | 107 | ||
X66117.1 | - | 2 - 3 | 434 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
