Assay Details
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111268.1 | NP_001104738.1 | 8 - 9 | 1506 | 95 | |
NM_010349.2 | NP_034479.2 | 8 - 9 | 1506 | 95 | ||
GenBank mRNA | BC131639.1 | - | 7 - 8 | 1187 | 95 | |
BC131640.1 | - | 7 - 8 | 1187 | 95 | ||
BC148311.2 | - | 7 - 8 | 1187 | 95 | ||
D10054.1 | - | 8 - 9 | 1419 | 95 | ||
X66117.1 | - | 7 - 8 | 1243 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
