Assay Details
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285872.1 | NP_001272801.1 | 6 - 7 | 708 | 88 | |
NM_001285874.1 | NP_001272803.1 | 5 - 6 | 650 | 88 | ||
NM_001285875.1 | NP_001272804.1 | 5 - 6 | 600 | 88 | ||
NM_001285877.1 | NP_001272806.1 | 3 - 4 | 344 | 88 | ||
NM_133880.3 | NP_598641.2 | 6 - 7 | 697 | 88 | ||
GenBank mRNA | AK144090.1 | - | 6 - 7 | 88 | ||
AK145092.1 | - | 5 - 6 | 551 | 88 | ||
AK156774.1 | - | 5 - 6 | 651 | 88 | ||
BC021890.1 | - | 5 - 6 | 602 | 3598000 | 88 | |
BC025495.1 | - | 6 - 7 | 719 | 5310200 | 88 | |
BC025938.1 | - | 5 - 6 | 543 | 88 | ||
BQ179290.1 | - | 4 - 5 | 419 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
