Assay Details
Gene Symbol: Sema4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB021291.1 | - | 6 - 7 | 769 | 67 | |
AF038652.1 | - | 5 - 6 | 528 | 67 | ||
AK035993.1 | - | 6 - 7 | 801 | 67 | ||
AK047067.1 | - | 6 - 7 | 785 | 67 | ||
AK143196.1 | - | 6 - 7 | 813 | 67 | ||
AK158292.1 | - | 6 - 7 | 735 | 67 | ||
BC057048.1 | - | 5 - 6 | 677 | 67 | ||
BC090625.1 | - | 6 - 7 | 771 | 67 | ||
BC094567.1 | - | 6 - 7 | 796 | 6834138 | 67 | |
BC139232.1 | - | 6 - 7 | 743 | 8862254 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
