Assay Details
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042743.2 | NP_001036208.1 | 21 - 22 | 2731 | 96 | |
NM_008641.3 | NP_032667.2 | 20 - 21 | 2710 | 96 | ||
GenBank mRNA | AB093264.1 | - | 11 - 12 | 1363 | 96 | |
AK004728.1 | - | 2 - 3 | 220 | 96 | ||
BC060703.1 | - | 21 - 22 | 2301 | 6412811 | 96 | |
CF743521.1 | - | 1 - 2 | 68 | 96 | ||
U02313.1 | - | 20 - 21 | 2380 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
