Assay Details
Gene Symbol: Cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026560.4 | NP_080836.3 | 8 - 9 | 973 | 89 | |
GenBank mRNA | AK003755.1 | - | 1 - 2 | 119 | 89 | |
AK076065.1 | - | 8 - 9 | 930 | 89 | ||
AK076422.1 | - | 8 - 9 | 963 | 89 | ||
AK077717.1 | - | 8 - 9 | 926 | 89 | ||
AK077920.1 | - | 8 - 9 | 896 | 89 | ||
AK078404.1 | - | 8 - 9 | 926 | 89 | ||
AK082800.1 | - | 8 - 9 | 938 | 89 | ||
AY550907.1 | - | 8 - 9 | 963 | 89 | ||
BC068181.1 | - | 7 - 8 | 887 | 30634239 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271160 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271162 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271159 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271158 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271161 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271163 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170681 | AY550907.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170681 | BC068181.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170681 | AK077717.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170681 | AK077920.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170681 | AK078404.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155142 | AK082800.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155142 | AK082800.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451651 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489736 | XM_894554.2 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489736 | XM_906003.2 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810422 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439343 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652104 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900831 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822433 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592567 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62602 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37214 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159286 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110084 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155142 | AK082800.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155142 | AK082800.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170681 | AY550907.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170681 | BC068181.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170681 | AK077717.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170681 | AK077920.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170681 | AK078404.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155142 | AK082800.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155142 | AK082800.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271158 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271154 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23271163 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159286 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00978661 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110084 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05388209 | - | - |
Whole Mouse Genome G4122A | A_51_P155142 | AK082800.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P155142 | AK082800.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P392544 | AY550907.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P392544 | BC068181.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P392544 | AK077717.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P392544 | AK077920.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P392544 | AK078404.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P463220 | AK031571.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138667 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120688 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159286 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110084 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03602283 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03602280 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03602278 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03602288 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03602289 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271160 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271159 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271158 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271162 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271163 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23271161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23270989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23271072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03915881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04098169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271085 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03953965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04098174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03602283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271150 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03431126 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271154 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03915883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03574877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03647985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271131 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271123 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23271139 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10516250 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516252 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516249 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516247 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516253 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516257 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516258 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516254 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516255 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516256 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516251 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428481_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428480_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436847_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10516246 | - | - |
Mouse Genome 430 2.0 Array | 1428480_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1428480_at | - | - |
Mouse Genome 430 2.0 Array | 1428481_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1436847_s_at | - | - |
Mouse Genome 430 2.0 Array | 1436847_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1428481_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52276_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52276_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400028536.mm.1 | TC0400003516.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400028536.mm.1 | NM_026560.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028536.mm.1 | NM_026560.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028533.mm.1 | TC0400003516.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400028533.mm.1 | NM_026560.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028533.mm.1 | NM_026560.4 | Refseq |
Mouse Expression Set 430 | 1436847_s_at | - | - |
Mouse Expression Set 430 | 1428481_s_at | - | - |
Mouse Expression Set 430 | 1428481_PM_s_at | - | - |
Mouse Expression Set 430 | 1428480_at | - | - |
Mouse Expression Set 430 | 1436847_PM_s_at | - | - |
Mouse Expression Set 430 | 1428480_PM_at | - | - |
Murine Genome U74v2 Set | 97295_PM_at | - | - |
Murine Genome U74v2 Set | 97295_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428481_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428480_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436847_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436847_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428480_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428481_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6980019 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2340286 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006980019 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002340286 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248269 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588362 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248269 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2588362 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2543393 | - | - |
mouse-6 v1.1 (Illumina) | scl0052276.1_330-S | - | - |
mouse-6 v1.1 (Illumina) | scl052276.4_53-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2340286 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6980019 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0052276.1_330-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl052276.4_53-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340286 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980019 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2588362 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2588362 | - | - |
