Assay Details
Gene Symbol: Sf3a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029157.3 | NP_083433.1 | 13 - 14 | 1203 | 96 | |
GenBank mRNA | AK030743.1 | - | 13 - 14 | 1238 | 96 | |
AK150733.1 | - | 13 - 14 | 1201 | 96 | ||
AK151141.1 | - | 13 - 14 | 1201 | 96 | ||
AK151382.1 | - | 13 - 14 | 1201 | 96 | ||
AK161646.1 | - | 13 - 14 | 1224 | 96 | ||
AK168966.1 | - | 13 - 14 | 1227 | 96 | ||
AK169324.1 | - | 13 - 14 | 1247 | 96 | ||
BC009141.1 | - | 13 - 14 | 1203 | 3598647 | 96 | |
BC092058.1 | - | 13 - 14 | 1235 | 4017388 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
