Assay Details
Gene Symbol: Lrig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008377.2 | NP_032403.2 | 13 - 14 | 2256 | 73 | |
GenBank mRNA | BC141300.1 | - | 13 - 14 | 2048 | 9055912 | 73 |
BC145623.1 | - | 13 - 14 | 2048 | 9054191 | 73 | |
D78572.1 | - | 13 - 14 | 2249 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
