Assay Details
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017383.3 | NP_059079.2 | 12 - 13 | 1805 | 102 | |
GenBank mRNA | AB032602.1 | - | 11 - 12 | 1491 | 102 | |
AK052972.1 | - | 13 - 14 | 1698 | 102 | ||
BC076594.1 | - | 12 - 13 | 1805 | 30649661 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
