Assay Details
Gene Symbol: Nr2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011630.3 | NP_035760.1 | 11 - 12 | 1635 | 85 | |
GenBank mRNA | AK157305.1 | - | 11 - 12 | 1609 | 85 | |
AK220507.1 | - | 4 - 5 | 533 | 85 | ||
BC103683.1 | - | 9 - 10 | 1254 | 40047943 | 85 | |
BC103684.1 | - | 9 - 10 | 1254 | 40047942 | 85 | |
BC103686.1 | - | 9 - 10 | 1254 | 40047941 | 85 | |
S75970.1 | - | 11 - 12 | 1750 | 85 | ||
U11688.1 | - | 10 - 11 | 1297 | 85 | ||
U32939.1 | - | 9 - 10 | 1268 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
