Assay Details
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081111.2 | NP_001074580.1 | 6 - 7 | 1918 | 114 | |
GenBank mRNA | BC150931.1 | - | 6 - 7 | 1929 | 9087842 | 114 |
BQ561257.1 | - | 1 - 2 | 128 | 114 | ||
CN456538.1 | - | 4 - 5 | 461 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
