Assay Details
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080448.4 | NP_536696.4 | 4 - 5 | 1260 | 81 | |
GenBank mRNA | AF338472.1 | - | 3 - 4 | 418 | 81 | |
AF496547.1 | - | 4 - 5 | 490 | 81 | ||
AK045508.1 | - | 4 - 5 | 1261 | 81 | ||
AK051262.1 | - | 4 - 5 | 1261 | 81 | ||
AK122276.1 | - | 4 - 5 | 908 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
