Assay Details
Gene Symbol: Vamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080742.2 | NP_001074211.1 | 1 - 2 | 91 | 66 | |
NM_016872.4 | NP_058568.1 | 1 - 2 | 91 | 66 | ||
GenBank mRNA | AK009266.1 | - | 1 - 2 | 94 | 66 | |
BC086925.1 | - | 1 - 2 | 45 | 6758831 | 66 | |
BY709062.1 | - | 1 - 2 | 88 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK009266.1, BC086925.1, NM_001080742.2, NM_016872.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
